Order Entry
Italy
ContactUsLinkComponent
Pesticide reference standard, Dicofol (2,2,2-Trichloro-1,1-bis(4-chlorophenyl)ethanol) 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Pesticide reference standard, Dicofol (2,2,2-Trichloro-1,1-bis(4-chlorophenyl)ethanol) 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERXA12570000CY
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, Dicofol (2,2,2-Trichloro-1,1-bis(4-chlorophenyl)ethanol) 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERXA12570000CY
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    115-32-2
  • Description:
    Dicofol (2,2,2-Trichloro-1,1-bis(4-chlorophenyl)ethanol) 100 µg/ml in cyclohexane
  • Formula:
    C₁₄H₉Cl₅O
  • Description:
    Dicofol (2,2,2-Trichloro-1,1-bis(4-chlorophenyl)ethanol)

Specifications

About this item