Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Desethyl atrazine
Desethyl atrazine
Desethyl atrazine
Desethyl atrazine
Catalog # EHERC10331000
Supplier:
EHRENSTORFER
CAS Number:
Expand image
Desethyl atrazine
Catalog # EHERC10331000
Supplier:
EHRENSTORFER
CAS Number:
Specifications
Pack type:
Glass bottle for solids
Pk:
0,25 g
Seq:
3029
Specifications
About this item
Product Details
:
:
215.69 g/mol
CAS Number:
6190-65-4
Formula:
C₈H₁₄ClN₅
Product Details