Diethyl isopropylmalonate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:759-36-4
:
Formula:C₁₀H₁₈O₄
MDL Number:MFCD00040491
:202.25 g/mol
Boiling Pt:214°C
Flash Pt:92°C
EINECS:212-068-0
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together