Order Entry
Italy
ContactUsLinkComponent
Pesticide reference standard, Aldrin 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Pesticide reference standard, Aldrin 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL10090000AL
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, Aldrin 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL10090000AL
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    309-00-2
  • Description:
    Aldrin 10 µg/ml in acetonitrile
  • Formula:
    C₁₂H₈Cl₆
  • Description:
    Aldrin

Specifications

About this item