Order Entry
Italy
ContactUsLinkComponent
 
Chromium standard solution, 10 mg/L Cr in 5% nitric acid (from Cr(NO₃)₃), standard for ICP, Specpure®
Catalog # 45251.AE
CAS Number:  
Chromium standard solution, 10 mg/L Cr in 5% nitric acid (from Cr(NO₃)₃), standard for ICP, Specpure®
Catalog # 45251.AE
CAS Number:  

Specifications

  • Concentration:
    10 mg/L
  • Matrix:
    5% HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • MDL No.:
    MFCD00010944
  • Element:
    Chromium
  • Symbol:
    Cr

Specifications

About this item

Molecular Formula: Cr(NO₃)₃ in 2% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010944

Certifications: Traceable to NIST