(3-Bromophenyl)acetone 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:21906-32-1
:
Formula:C₉H₉BrO
MDL Number:MFCD00210400
:213.07 g/mol
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear light yellow to dark yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5570 to 1.5600 (20°C, 589 nm)
- GC:≥98.0 %
Specification Test Results
Frequently Bought Together