
2,6-Di-tert-butylpyridine ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:585-48-8
:
Formula:C₁₃H₂₁N
MDL Number:MFCD00006306
:191.32 g/mol
Merck Index:14,03036
Boiling Pt:101°C
Melting Pt:2°C
Density:0,852 g/cm³ (25 °C)
Flash Pt:71°C
EINECS:209-557-6
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together