

1,8-Dinitronaphthalene ≥85.0% (by GC)
Catalog # TCIAD0837-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (GC):min. 85.0 %
- Melting point:150.0 to 172.0 deg-C
- Solubility in hot Dioxane:almost transparency
Specification Test Results
About this item
:
:218.17 g/mol
Boiling Pt:390 °C (1013 hPa)
Melting Pt:173 °C
EINECS:210-016-1
:GHS07
:GHS08
CAS Number:602-38-0
Formula:C₁₀H₆N₂O₄
MDL Number:MFCD00003918