Laboratory glass cleaner, borer 22 HPF-x
Supplier: Borer Chemie
Certificates
About this item
Liquid, highly alkaline cleaning concentrate for automated cleaning of laboratory glassware und reusable laboratory material.
- Rinsable without residues and guarantees therefore consistent and error-free results
- With a broad cleaning spectrum and excellent results even at low concentrations
- High cleaning performance due to excellent dispersing capabilities, effective against stubborn deposits
- Well suited for an effective cleaning of animal cages
- Phosphate- and surfactant-free, no release of phosphates and surfactants into waste water
- EDTA-, NTA- and chlorine-free, environmentally friendly
- Rinsing with an acidic deconex® cleaner is recommended as the completion of the cleaning process
Frequently Bought Together