Laboratory glass cleaner, borer 15 NF
Supplier: Borer Chemie
Certificates
About this item
Liquid, alkaline cleaning concentrate for manual cleaning of laboratory glassware and reusable laboratory materials. Is especially suitable for the pre-cleaning before automated cleaning - also without intermediate rinsing.
- Rinsable without residues and guarantees therefore consistent and error-free results
- With a broad cleaning spectrum and perfect results even at low concentrations
- Working solution suitable for multiple use
- Facilitates the complete detachment of stubborn deposits in automated cleaning
- Residues carried over from pre-cleaning do not cause foaming in laboratory washers
- Phosphate-free, suitable for phosphate-critical applications
- EDTA-, NTA- and chlorine-free, environmentally friendly
Suitable for soaking baths, pipette washers and ultrasonic devices.
Frequently Bought Together