
Adipoyl dichloride for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:111-50-2
:
Formula:C₆H₈Cl₂O₂
MDL Number:MFCD00000759
:183.03 g/mol
Boiling Pt:125 °C (15 hPa)
Density:1.2563 g/cm³ (20 °C)
Flash Pt:66 °C
EINECS:203-876-4
ADR:8,II
:M-Clarity™ quality level = MQ200
UN:3265
:GHS05
Specification Test Results
- Density:1.259 g/cm3 (25 °C)
Specification Test Results
Frequently Bought Together