Order Entry
Italy
ContactUsLinkComponent
Ammonium standard solution, 1000 mg/L NH₄ in water (from (NH₄)₂SO₄), standard for Ion Chromatography, Specpure®
Ammonium standard solution, 1000 mg/L NH₄ in water (from (NH₄)₂SO₄), standard for Ion Chromatography, Specpure®
Catalog # 35560.AE
CAS Number:  
Ammonium standard solution, 1000 mg/L NH₄ in water (from (NH₄)₂SO₄), standard for Ion Chromatography, Specpure®
Catalog # 35560.AE
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Description:
    Ammonium standard solution
  • MDL No.:
    MFCD00143604
  • Contents:
    (NH₄)₂SO₄ in H₂O

Specifications

About this item

Standard from (NH₄)₂SO₄ in H₂O

Certifications: Traceable to NIST