Isopropyl myristate ≥98.0% (by GC)
: TCI
:110-27-0
:
:C₁₇H₃₄O₂
:MFCD00008982
:270.46 g/mol
:14,05215
:193°C
:0…1 °C
:0,852 g/cm³ (20 °C)
:164°C
:Refrigerator
:203-751-4
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Frequently Bought Together