Dip culture media for bacteriological urine analysis, Cult-Dip® plus
Catalog # 1.00777.0001
Supplier: Merck
CAS Number:
Specifications
- Pk:10
- Description:Cult-Dip® plus (Brolacin/MacConkey-Agar)
Specifications
About this item
Cult-Dip® plus is a bacteriological culture test for detecting and enumerating E. coli in urine samples. Fluorescent detection using a UV lamp enables E. coli infections to be clearly identified.
- Allows multiple isolates of E.coli to be identified
- Simple handling
- Quick and clear analysis