Order Entry
Italy
Orders LinkContactUsLinkComponent
Hypodermic safety needles, Needle-Pro®
Hypodermic safety needles, Needle-Pro®
Catalog #
Hypodermic safety needles, Needle-Pro®
Catalog #
Supplier:  
Hypodermic safety needles, Needle-Pro®
Hypodermic safety needles, Needle-Pro®
Product Details & Documents
Stainless steel cannula, PP sheath and hub. The hypodermic Needle-Pro® devices are specifically designed to minimise the risk of needlestick injuries. Visual and audible confirmation as the needle locks into place with a simple one-handed technique that keeps the user’s hand behind the needle at all times.
  • Designed to allow standard puncture technique without requiring extra steps
  • Luer slip 26 to 30G devices compatible with Luer slip syringes
  • Luer lock and Luer slip 18 to 25G devices compatible with both Luer slip and Luer lock syringes
Certifications: In accordance with ISO 7864. Colour-coded hub conforms to ISO 6009.