Temperature probes, Lemo®, BOLA
Supplier: Bohlender
Certificates
About this item
Used for temperature measurement in aggressive liquids and cable provides flexible connection from measuring device to medium.
- Thermocouple in a PTFE-encapsulated stainless steel tube
- Temperature probe of tip Ø 6mm and collar ring Ø 12mm
- With white PTFE-coated cable (length of 1,5 m) and coupling (type Lemo® socket size 1, 4-pole)
- Typical response times of T50 is 7 to 12 seconds and T90 is 14 to 16 seconds
Frequently Bought Together