Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Magnesia
Magnesia, troughs, Length: ±100 mm, Width: 9 mm
Magnesia, troughs, Length: ±100 mm, Width: 9 mm
Magnesia, troughs, Length: ±100 mm, Width: 9 mm
Catalog # 459-2911
Nome del fornitore:
CERAM TEC
Numero CAS:
Expand image
Magnesia, troughs, Length: ±100 mm, Width: 9 mm
Catalog # 459-2911
Nome del fornitore:
CERAM TEC
Numero CAS:
Specifiche
Pk:
100
Seq:
0100
Specifiche
Informazioni su questo articolo
Product Details
:
:
1309-48-4
Product Details