SP Bel-Art bench-top biohazard bags
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Bright red, autoclavable bags feature one long flap for easy opening. Printed with the biohazard symbol and sterilization indicator patch, they are made of High Molecular Weight, High-Density (HMHD) polyethylene.
- Use as a biohazard material handler: Small size allows the bag to be inverted over your hand to safely pick up waste
- Steam autoclavable at 121 °C
For a space-saving disposal unit in fume hoods or on bench tops, use with Poxygrid® Benchtop Biohazard Bag Holder (129-4948) and Cover (129-1475).
Bag holder kit and components
Product Information
Frequently Bought Together