6,7-Dimethoxyquinazoline-2,4(1H,3H)-dione
Catalog # PROOMM0085.07
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:222.2 g/mol
EINECS:249-288-1
CAS Number:28888-44-0
Formula:C₁₀H₁₀N₂O₄
MDL Number:MFCD00023889