Order Entry
Italy
ContactUsLinkComponent
4,4-Dimethyl-2-oxazoline for synthesis, Sigma-Aldrich®
Flammable
4,4-Dimethyl-2-oxazoline for synthesis, Sigma-Aldrich®
Catalog # 8.18107.0005
Supplier:  Merck
CAS Number:  
Flammable
4,4-Dimethyl-2-oxazoline for synthesis, Sigma-Aldrich®
Catalog # 8.18107.0005
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • Solubility:
    (20 °C) (decomposition)
  • Molar Mass:
    99.13 g/mol
  • Boiling Point:
    99 - 100 °C (1013 hPa)
  • Flash Point:
    1 °C
  • Density:
    0.94 g/cm3 (20 °C)

Specification Test Results

About this item

 :
 :99.13 g/mol
ADR:3,II
 :M-Clarity™ quality level = MQ200
EINECS:250-045-7
UN:1993
 :GHS02
Storage Temperature:Ambient
CAS Number:30093-99-3
Formula:C₅H₉NO
MDL Number:MFCD00014507