




tert-Butyl hydroperoxide ≥70% in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75-91-2
:
Formula:C₄H₁₀O₂
MDL Number:MFCD00002130
:90.12 g/mol
Merck Index:14,1570
Boiling Pt:96 °C (dec.)
Melting Pt:–3 °C
Density:0.937
Flash Pt:43 °C(109 °F)
EINECS:200-915-7
UN:3109
ADR:5.2,
:GHS02
:GHS05
:GHS06
:GHS08
:GHS09
Storage Temperature:Refrigerator
Related Information
Frequently Bought Together