
Potassium-2,5-dihydroxybenzenesulphonate for synthesis
: Merck
:21799-87-1
:
:C₆H₅KO₅S
:MFCD00007475
:228.27 g/mol
:M-Clarity™ quality level = MQ200
:244-584-7
:GHS07
:Ambient
- Solubility:(20 °C) soluble
- Melting Point:250 - 251 °C
- Molar Mass:228.27 g/mol
Frequently Bought Together