Order Entry
Italy
ContactUsLinkComponent
 
 
 
 
Trimellitic anhydride for synthesis, Sigma-Aldrich®
 :  Merck
 
 
 
 
Trimellitic anhydride for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Solubility:
    (20 °C) (decomposition)
  • Melting Point:
    164 - 168 °C
  • Molar Mass:
    192.12 g/mol
  • Boiling Point:
    240 °C (19 hPa)
  • Vapor Pressure:
    19 hPa (240 °C)
  • Flash Point:
    227 °C
  • Explosion Limit:
    1 - 7 %(V)
  • pH:
    2 (21 g/l, H2O, 20 °C)

 

 

Frequently Bought Together