Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Succinylsulfathiazole
Succinylsulphathiazole
Succinylsulphathiazole
Succinylsulphathiazole
EHERC16985600
:
EHRENSTORFER
:
Succinylsulphathiazole
EHERC16985600
:
EHRENSTORFER
:
Pack type:
Glass bottle for solids
Pk:
0,25 g
Seq:
3029
:
:
355.38 g/mol
:
204-141-0
:
116-43-8
:
C₁₃H₁₃N₃O₅S₂
:
MFCD00022437