
2,6-Di-tert-butyl-p-benzoquinone ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:719-22-2
:
Formula:C₁₄H₂₀O₂
MDL Number:MFCD00001601
:220.31 g/mol
Melting Pt:65…69 °C
Flash Pt:>110 °C (230 °F)
EINECS:211-946-0
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together