Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Phenylbutazone
Phenylbutazone
Phenylbutazone
Phenylbutazone
EHERC16056500
:
EHRENSTORFER
:
Phenylbutazone
EHERC16056500
:
EHRENSTORFER
:
Pack type:
Glass bottle for solids
Pk:
0,1 g
Seq:
3020
:
:
308.38 g/mol
:
200-029-0
:
2811
:
6.1,III
:
50-33-9
:
C₁₉H₂₀N₂O₂
:
MFCD00005500