Ammonium standard solution, 200 mg/L NH₄ in water standard for Ion Chromatography
Catalog # REGEICCT01
Supplier: REAGECON
CAS Number:
Specifications
- Concentration:200 mg/L
- Matrix:H₂O
- Pack type:Glass bottle
- Pk:100 mL
- Description:Ammonium standard solution