Barium standard solution, 1,000 mg/L Ba in dil. nitric acid (from BaCO₃) standard for AAS and ICP
Catalog # PERCN9300181
Nome del fornitore: PerkinElmer
Numero CAS:
Specifiche
- Concentration:1000 mg/L
- Matrix:2% HNO₃
- Pack type:Plastic bottle
- Pk:125 mL
- Element:Barium
- Symbol:Ba