Hand protectors, Hot Grip
Catalog # KART3915
Supplier: KARTELL
Specifications
- Colour:Red
- Pk:1
Specifications
About this item
These hand protectors made of silicone rubber provide protection against hot containers up to +260 °C and cold containers down to –57 °C. Internal relief surface.