Silicone oil, 100 cSt 47 V 100
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:63148-62-9
MDL Number:MFCD00132673
Density:0.965 g/cm³ (20 °C)
Flash Pt:321 °C (closed cup)
Storage Temperature:Ambient
Specification Test Results
- Recommended working temperature:-5 to +200°C
- Viscosity (25°C):92 - 108 cSt
Specification Test Results
Related Information
Certificates
Search by Lot Number Frequently Bought Together