Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Iodine monobromide
Iodine monobromide 20 g/l in acetic acid (99%) Reag. Ph. Eur. 1043504
Iodine monobromide 20 g/l in acetic acid (99%) Reag. Ph. Eur. 1043504
REGE1045901
:
REAGECON
:
Iodine monobromide 20 g/l in acetic acid (99%) Reag. Ph. Eur. 1043504
REGE1045901
:
REAGECON
:
Pack type:
Glass bottle
Pk:
1 L
Seq:
5037
:
:
206.81 g/mol
:
Ambient
:
232-159-9
:
2789
:
8,II
:
GHS02
:
GHS05
:
7789-33-5
:
IBr
:
MFCD00011353