GL tube fittings, PTFE EX
Supplier: Bohlender
Certificates
About this item
Connector for tubing, made of PTFE-EX, with two GL screw connections. To distribute liquids or gases. Can be used as a reducer to connect different tubes or hoses.
- Very good chemical resistance
- Temperature resistant from −200 to +250 °C
- Pressure resistant up to a maximum of 10 bar, vacuum suitable
Frequently Bought Together