Order Entry
Italy
Orders LinkContactUsLinkComponent
Test strips, sulfite, QUANTOFIX®

Test strips, sulfite, QUANTOFIX®

QUANTOFIX Sulfite, 0 • 10 • 25 • 50 • 100 • 250 • 500 • 1000 - mg/L - SO₃²⁻ ; 10–500 - mg/L - SO₃²⁻, Colour reaction with sodiumnitroprusside, potassium hexacyanoferrat, zinc sulfate, 40×123×40 mm (package), 100 test(s), 100 Tests
Catalog # 705-0307
Supplier:   MACHEREY-NAGEL
Test strips, sulfite, QUANTOFIX®
Catalog # 705-0307
Supplier:  MACHEREY-NAGEL
CAS Number:  
Test strips, sulfite, QUANTOFIX®
Test strips, sulfite, QUANTOFIX®

QUANTOFIX Sulfite, 0 • 10 • 25 • 50 • 100 • 250 • 500 • 1000 - mg/L - SO₃²⁻ ; 10–500 - mg/L - SO₃²⁻, Colour reaction with sodiumnitroprusside, potassium hexacyanoferrat, zinc sulfate, 40×123×40 mm (package), 100 test(s), 100 Tests

CAS Number:  
Price
34,30 €
Log in for availability
1 * 100 Tests
Product Details & Documents

QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. These test strips are immediately ready-to-use. They do not require additional accessories. The test strips are intended for single use, maintenance or calibration are not required.

  • Results within seconds: Just dip, wait and read against multi-stage colour scale
  • Always ready to use: No accessories necessary for evaluation
  • Safe results: Desiccant in the stopper for optimal protection of the strips against humidity

These test strips are for the rapid and reliable determination of Sulphite in solutions. The easy dip-and-read-procedure provides a reliable result within 20 seconds.

In process and boiler water, Sulphite is used as an oxygen scavenger. To avoid over treatment, the concentration needs to be controlled, regularly. These tests are also used to control the Sulphite concentration in foodstuff treated with Sulfur compounds (SO₂, HSO₃⁻, SO₃²⁻) for longer shelf-life. In the process of wine making the control of Sulphite is important to control production and quality of the wine.

Gradation: 0 · 10 · 25 · 50 · 100 · 250 · 500 · 1000 mg/L SO₃²⁻

Colour change: white → salmon

Specifications

Specifications

  • Chemical method:
    Colour reaction with sodiumnitroprusside, potassium hexacyanoferrat, zinc sulfate
  • Pk:
    100 Tests
  • Description:
    QUANTOFIX Sulfite
  • Dimensions:
    40×123×40 mm (package)
  • Tests per kit:
    100 test(s)
  • Range:
    0 • 10 • 25 • 50 • 100 • 250 • 500 • 1000 - mg/L - SO₃²⁻ ; 10–500 - mg/L - SO₃²⁻
  • Cat. No.:
    705-0307
  • Parameters:
    All reagents included
  • Contents:
    100 test strips in a tube, instruction leaflet
  • Platform:
    Semi-quantitative test strips
  • Compliance:
    No hazardous material
  • Instrument:
    QUANTOFIX Relax
  • Shelf life (months):
    30 (from production)
  • Method:
    Sulfite - Nitroprussid / Zn-hexacyanoferrate (white → salmon)
  • Weight:
    42 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.