Order Entry
Italy
ContactUsLinkComponent
Benzo-18-crown-6 (benzo-1,4,7,10,13,16-hexoxacyclooctadecane) 97%
Irritant
Benzo-18-crown-6 (benzo-1,4,7,10,13,16-hexoxacyclooctadecane) 97%
Certificates
undefined
Irritant
Benzo-18-crown-6 (benzo-1,4,7,10,13,16-hexoxacyclooctadecane) 97%

Specification Test Results

  • Appearance:
    White to off-white Powder or crystals and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    40 to 45 °C
  • GC:
    ≥96.0 %

Specification Test Results

Related Information

Frequently Bought Together