

6,9-Dichloro-2-methoxyacridine ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:86-38-4
:
Formula:C₁₄H₉Cl₂NO
MDL Number:MFCD00005028
Melting Pt:163…165 °C
:278.14 g/mol
EINECS:201-666-7
:GHS08
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together