

Cyclopentyl methyl ether (CPME) ≥99.9%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5614-37-9
:
Formula:C₆H₁₂O
MDL Number:MFCD08276401
:100.16 g/mol
Boiling Pt:106 °C
Melting Pt:–140 °C
Density:0.86
Flash Pt:–1 °C (–30 °F)
Storage Temperature:Ambient
UN:3271
ADR:3,II
:GHS02
:GHS07
Related Information
Frequently Bought Together