




Hydroxylamine 50% in aqueous solution for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7803-49-8
:
Formula:H₃NO
MDL Number:MFCD00044522
:33.03 g/mol
Density:1.12 g/cm3 (20 °C)
Storage Temperature:Ambient
EINECS:232-259-2
UN:3266
ADR:8,III
:M-Clarity™ quality level = MQ200
:GHS02
:GHS05
:GHS07
:GHS08
:GHS09
Specification Test Results
- Assay (manganometric):47.0 - 53.0 %
- Density (d 20 °C/ 4 °C):1.120 - 1.123
- Vapor Pressure:14 hPa (20 °C)
- Density:1.12 g/cm3 (20 °C)
- pH:10.6 (H2O, 20 °C)
- Ignition Point:215 °C
- Viscosity kinematic:1.435 mm2/s (25 °C)
- Hazard Class:9
- Hazard Class Description:Miscellaneous hazardous material
Specification Test Results
Related Information
Frequently Bought Together