Organic reference standard, 2,2-Dichloropropionic acid 1000 µg/ml in tert-butyl methyl ether, Dr Ehrenstorfer
EHERYA11949500MB
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:75-99-0
- Description:2,2-Dichloropropionic acid 1,000 µg/ml in tert-butyl methyl ether
- Formula:C₃H₄Cl₂O₂
- Description:2,2-Dichloropropionic acid