1-Chloroethylethyl carbonate for synthesis
Catalog # 8.10847.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 ml
- Seq:2440
Specifications
About this item
:
:152.58 g/mol
EINECS:256-832-1
CAS Number:50893-36-2
Formula:C₅H₉ClO₃
MDL Number:MFCD00040523