Order Entry
Italy
ContactUsLinkComponent
Organic reference standard, 1-Hexacosanol 10 µg/ml in tert-butyl methyl ether, Dr Ehrenstorfer
Organic reference standard, 1-Hexacosanol 10 µg/ml in tert-butyl methyl ether, Dr Ehrenstorfer
Codice catalogo EHERL14191400MB
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, 1-Hexacosanol 10 µg/ml in tert-butyl methyl ether, Dr Ehrenstorfer
Codice catalogo EHERL14191400MB
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    506-52-5
  • Description:
    1-Hexacosanol 10 µg/ml in tert-butyl methyl ether
  • Formula:
    C₂₆H₅₄O
  • Description:
    1-Hexacosanol

Specifications

About this item