Order Entry
Italy
ContactUsLinkComponent
Organic reference standard, Chloroacetic acid 1000 µg/ml in tert-butyl methyl ether, Dr Ehrenstorfer
Organic reference standard, Chloroacetic acid 1000 µg/ml in tert-butyl methyl ether, Dr Ehrenstorfer
Catalog # EHERYA11348500MB
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, Chloroacetic acid 1000 µg/ml in tert-butyl methyl ether, Dr Ehrenstorfer
Catalog # EHERYA11348500MB
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    79-11-8
  • Description:
    Chloroacetic acid 1,000 µg/ml in tert-butyl methyl ether
  • Formula:
    C₂H₃ClO₂
  • Description:
    Chloroacetic acid

Specifications

About this item