


Thiophene for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:110-02-1
:
Formula:C₄H₄S
MDL Number:MFCD00005413
:84.14 g/mol
Merck Index:12,09490
Boiling Pt:84.4 °C (1013 hPa)
Melting Pt:–38 °C
Density:1.06 g/cm³ (20 °C)
EINECS:203-729-4
UN:2414
ADR:3,II
:M-Clarity™ quality level = MQ200
Flash Pt:–9 °C
:GHS02
:GHS07
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:-38 °C
- Molar Mass:84.14 g/mol
- Boiling Point:83 - 84 °C (1013 mbar)
- Vapor Pressure:80 hPa (20 °C)
- Flash Point:-9 °C
- Refractive Index:1.5289 (20 °C, 589 nm)
- Explosion Limit:1.5 - 12.5 %(V)
- Density:1.06 g/cm3 (20 °C)
- Ignition Point:395 °C
- Saturation concentration:276 g/m3 (20 °C) Air
- Hazard Class:3
- Hazard Class Description:Flammable Liquid
Specification Test Results
Frequently Bought Together