
2-Iodothiophene 98+%
ACRO187880100
:
- Pack type:Glass bottle
- Pk:10 mL
- Seq:2275
- Appearance:Clear yellow to red-brown to blue-green Liquid
- Infrared spectrum:Conforms
- Refractive index:1.6500 to 1.6540 (20°C, 589 nm)
- GC:≥98.0 %
:
:210.04 g/mol
:182 °C (1013 hPa)
:–40 °C
:1.9 g/cm³
:71 °C
:Refrigerator
:222-342-1
:GHS07
:3437-95-4
:C₄H₃IS
:MFCD00005424