


3-Bromothiophene 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:872-31-1
:
Formula:C₄H₃BrS
MDL Number:MFCD00005464
:163.04 g/mol
Boiling Pt:158 °C (760 mmHg)
Melting Pt:-10 °C
Density:1.74 g/cm³
Flash Pt:56 °C
Storage Temperature:Ambient
EINECS:212-821-3
UN:3384
ADR:6.1,
:GHS06
:GHS09
:GHS02
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid (May darken on storage)
- Infrared spectrum:Conforms
- Refractive index:1.5905 to 1.5925 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together