
3,4-Dibromothiophene 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3141-26-2
:
Formula:C₄H₂Br₂S
MDL Number:MFCD00005465
:241.93 g/mol
Boiling Pt:221…222 °C (760 mmHg)
Melting Pt:4…5 °C
Density:2.13 g/cm³
Flash Pt:>110 °C
Storage Temperature:Refrigerator
EINECS:221-546-8
UN:2810
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:Clear colorless to yellow to light brown Liquid (May darken on storage)
- Infrared spectrum:Conforms
- Refractive index:1.6380 to 1.6420 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together