Order Entry
Italy
ContactUsLinkComponent
 
 
 
2-Bromothiophene 98%
 
undefined
 
 
2-Bromothiophene 98%

 

  • Appearance:
    Clear colorless to light brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5850 to 1.5870 (20°C, 589 nm)
  • GC:
    ≥97.5 %
  • NMR (1H-NMR):
    Consistent with structure

 

 

Frequently Bought Together