
2,6-Diaminopyridine for synthesis, Sigma-Aldrich®
Catalog # 8.03575.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Solubility:180 g/l (20 °C)
- Melting Point:115 - 118 °C
- Molar Mass:109.13 g/mol
- Boiling Point:159 °C (13 hPa)
- Flash Point:155 °C
- pH:7 (H2O)
Specification Test Results
About this item
:
:109.13 g/mol
Boiling Pt:285 °C (1013 hPa)
Melting Pt:115…118 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,I
:M-Clarity™ quality level = MQ200
Flash Pt:155 °C
EINECS:205-507-2
:GHS06
CAS Number:141-86-6
Formula:C₅H₇N₃
MDL Number:MFCD00006329