Order Entry
Italy
Orders LinkContactUsLinkComponent
2,6-Difluoropyridine ≥98%

2,6-Difluoropyridine ≥98%

Flammable
Irritant
2,6-Difluoropyridine ≥98%
Certificates
2,6-Difluoropyridine ≥98%
2,6-Difluoropyridine ≥98%

3 Options Available Below

Product Details & Documents

Purity: 98+% CAS No.: 1513-65-1 Molecular Formula: C5H3F2N Formula Weight: 115.08 Flash Point: 33°(91°F) Refractive Index: 1.4370 Odour: Characteristic Boiling Point: 123-124° Beilstein Reference: 1422549 UN No.: 1993 MDL No.: MFCD00006227

  • CAS Number:1513-65-1
  • Formula:C₅H₃F₂N
  • MDL Number:MFCD00006227
  • Molecular Weight:115.08 g/mol
  • Boiling Pt:123…124 °C
  • Density:1.268
  • Flash Pt:33 °C (91 °F)
  • Storage Temperature:Ambient
  • EINECS:216-152-8
  • UN:1993
  • ADR:3,III
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.