Order Entry
Italy
ContactUsLinkComponent
Industrial gloves, Polytril™ Air Comfort
Industrial gloves, Polytril™ Air Comfort
Supplier:  Honeywell Safety
Certificates
undefined
Industrial gloves, Polytril™ Air Comfort
Supplier:  Honeywell Safety
Specifications
  • Catalog No:
  • 112-1541
  • 112-1537
  • 112-1539
  • 112-1540
  • 112-1538
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Cotton/Lycra®
  • Cotton/Lycra®
  • Cotton/Lycra®
  • Cotton/Lycra®
  • Cotton/Lycra®
  • Finish:
  • Palm and fingers coated
  • Palm and fingers coated
  • Palm and fingers coated
  • Palm and fingers coated
  • Palm and fingers coated
  • Gauge:
  • 13
  • 13
  • 13
  • 13
  • 13
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (3121X)
  • EN 388:2016 (3121X)
  • EN 388:2016 (3121X)
  • EN 388:2016 (3121X)
  • EN 388:2016 (3121X)