Order Entry
Italy
Orders LinkContactUsLinkComponent
 

1,3-Dimethylnaphthalene 96%

Glass bottle for solids, 1 g
Catalog # ACRO346900010
1,3-Dimethylnaphthalene 96%
Catalog # ACRO346900010
CAS Number:  575-41-7
1,3-Dimethylnaphthalene 96%
1,3-Dimethylnaphthalene 96%

Glass bottle for solids, 1 g

CAS Number:  575-41-7
Price
41,20 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:575-41-7
  • Formula:C₁₂H₁₂
  • MDL Number:MFCD00004036
  • Molecular Weight:156.23 g/mol
  • Boiling Pt:263 °C
  • Melting Pt:–5 °C
  • Density:0.982 g/cm³
  • Flash Pt:109 °C
  • Storage Temperature:Ambient
  • EINECS:209-384-6

 

Specifications

Specification Test Results

  • Appearance:
    Clear yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.6080 to 1.6100 (20°C, 589 nm)
  • GC:
    ≥95 %

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Cat. No.:
    ACRO346900010

Specifications